Product Details

SNP ID
rs78936999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16185575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCCCTTAGACCTTTTCTCGGGA[A/C]CCCTGGCCCTGATCCTCCGACCCGC
Phenotype
MIM: 614554
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM32A PubMed Links

Gene Details

Gene
FAM32A
Gene Name
family with sequence similarity 32 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014077.3 Intron NP_054796.1
XM_011527898.1 Intron XP_011526200.1

View Full Product Details