Product Details

SNP ID
rs77931469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16363903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAAAAACAGTCTTCCTTCCCAC[A/G]GTTTGCACAGAAGGACTCGGAGACA
Phenotype
MIM: 616826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPS15L1 PubMed Links

Gene Details

Gene
EPS15L1
Gene Name
epidermal growth factor receptor pathway substrate 15 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258374.1 Intron NP_001245303.1
NM_001258375.1 Intron NP_001245304.1
NM_001258376.1 Intron NP_001245305.1
NM_021235.2 Intron NP_067058.1
XM_017027086.1 Intron XP_016882575.1
XM_017027087.1 Intron XP_016882576.1
XM_017027088.1 Intron XP_016882577.1
XM_017027089.1 Intron XP_016882578.1
XM_017027090.1 Intron XP_016882579.1
XM_017027091.1 Intron XP_016882580.1
XM_017027092.1 Intron XP_016882581.1

View Full Product Details