Product Details

SNP ID
rs74372826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45124050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACTAATTCCTCCGAACTTTCAT[A/G]GGTCAGATAGAGGCAGGGAAGGGAC
Phenotype
MIM: 609872 MIM: 605161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WFDC12 PubMed Links

Gene Details

Gene
WFDC12
Gene Name
WAP four-disulfide core domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080869.1 Intron NP_543145.1
Gene
WFDC5
Gene Name
WAP four-disulfide core domain 5
There are no transcripts associated with this gene.

View Full Product Details