Product Details
- SNP ID
-
rs80339443
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:61256907 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGAAGAAAGAGATGTTTTATATC[C/T]TACAAATAATTTTGCACATATGGAG
- Phenotype
-
MIM: 603006
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH4
PubMed Links
Gene Details
- Gene
- CDH4
- Gene Name
- cadherin 4
View Full Product Details