Product Details

SNP ID
rs76972265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35709861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATCTAGCCCATAATGGGGCAAC[C/T]TGATTATTGATTTCCTGGCCTAGTG
Phenotype
MIM: 604739
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM39 PubMed Links

Gene Details

Gene
RBM39
Gene Name
RNA binding motif protein 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242599.1 Intron NP_001229528.1
NM_001242600.1 Intron NP_001229529.1
NM_001323422.1 Intron NP_001310351.1
NM_001323423.1 Intron NP_001310352.1
NM_001323424.1 Intron NP_001310353.1
NM_004902.3 Intron NP_004893.1
NM_184234.2 Intron NP_909122.1
XM_006723890.3 Intron XP_006723953.1
XM_006723891.3 Intron XP_006723954.1
XM_011529111.2 Intron XP_011527413.1
XM_017028138.1 Intron XP_016883627.1
XM_017028139.1 Intron XP_016883628.1
XM_017028140.1 Intron XP_016883629.1
XM_017028141.1 Intron XP_016883630.1
XM_017028142.1 Intron XP_016883631.1
XM_017028143.1 Intron XP_016883632.1
XM_017028144.1 Intron XP_016883633.1
XM_017028145.1 Intron XP_016883634.1
XM_017028146.1 Intron XP_016883635.1
XM_017028147.1 Intron XP_016883636.1
Gene
ROMO1
Gene Name
reactive oxygen species modulator 1
There are no transcripts associated with this gene.

View Full Product Details