Product Details

SNP ID
rs74351785
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18024832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATCCTCGCAGACGTAGAGCTTG[C/T]CCCGCCGCTGCTTATAGGCATACTG
Phenotype
MIM: 616441
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OVOL2 PubMed Links

Gene Details

Gene
OVOL2
Gene Name
ovo like zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303461.1 880 Missense Mutation GAC,GGC D79G NP_001290390.1
NM_001303462.1 880 Missense Mutation GAC,GGC D79G NP_001290391.1
NM_021220.3 880 Missense Mutation GAC,GGC D211G NP_067043.2

View Full Product Details