Product Details

SNP ID
rs76524831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:271216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATGGCACTGCCATCTAGAAGGG[C/T]AGTGGCTCTTCCACAGGAATGTTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C20orf96 PubMed Links

Gene Details

Gene
C20orf96
Gene Name
chromosome 20 open reading frame 96
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080571.1 1181 Silent Mutation CTA,CTG L360L NP_542138.1
NM_153269.2 1181 Silent Mutation CTA,CTG L361L NP_695001.2
XM_006723543.3 1181 Silent Mutation CTA,CTG L308L XP_006723606.1

View Full Product Details