Product Details

SNP ID
rs78896118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:274496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGCCTAATTCTGGGTTCACTAT[G/T]CAGAGGCCAACACAGTCTCTAAAAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C20orf96 PubMed Links

Gene Details

Gene
C20orf96
Gene Name
chromosome 20 open reading frame 96
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080571.1 Intron NP_542138.1
NM_153269.2 Intron NP_695001.2
XM_006723543.3 Intron XP_006723606.1

View Full Product Details