Product Details

SNP ID
rs79914010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3779315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGCGCGATCTTGCGCATCACGT[C/T]ATCCGGTACTGAAAAGTTCAGCCTC
Phenotype
MIM: 116949 MIM: 117140 MIM: 610674
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC25B PubMed Links

Gene Details

Gene
CDC25B
Gene Name
cell division cycle 25B
There are no transcripts associated with this gene.

Gene
CENPB
Gene Name
centromere protein B
There are no transcripts associated with this gene.

Gene
SPEF1
Gene Name
sperm flagellar 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015417.4 438 Missense Mutation AAC,GAC N87D NP_056232.2
XM_005260683.4 438 Missense Mutation AAC,GAC N87D XP_005260740.1

View Full Product Details