Product Details

SNP ID
rs76773684
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29197917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGATGGGAAAATAGAGGAGAGG[C/T]AGGAGAAAATGTGGGGCAGTGGGAA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LINC00189 PubMed Links

Gene Details

Gene
LINC00189
Gene Name
long intergenic non-protein coding RNA 189
There are no transcripts associated with this gene.

View Full Product Details