Product Details

SNP ID
rs74823159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30425632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACCCCGAGACCCAACATGCATA[A/G]TCAGGCAAGAATTGCAGAGCATGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP13-3 PubMed Links

Gene Details

Gene
KRTAP13-3
Gene Name
keratin associated protein 13-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181622.1 281 Missense Mutation ACT,ATT T94I NP_853653.1
Gene
KRTAP13-4
Gene Name
keratin associated protein 13-4
There are no transcripts associated with this gene.

View Full Product Details