Product Details

SNP ID
rs79258920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30881292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCAAGTCACCTGGCAAGGGTTG[A/G]AGACACATGAAGTTCGCCGGTAACA
Phenotype
MIM: 600064
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP11-1 PubMed Links

Gene Details

Gene
KRTAP11-1
Gene Name
keratin associated protein 11-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175858.2 264 Missense Mutation TCC,TTC S78F NP_787054.1

View Full Product Details