Product Details

SNP ID
rs79446227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38124317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTTAATGTCTCTGCATCAACT[C/T]GGGTTTCCTTTTGCTTTTGTTTAAA
Phenotype
MIM: 604829 MIM: 613396
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSCR4 PubMed Links

Gene Details

Gene
DSCR4
Gene Name
Down syndrome critical region 4
There are no transcripts associated with this gene.

Gene
DSCR8
Gene Name
Down syndrome critical region 8
There are no transcripts associated with this gene.

View Full Product Details