Product Details

SNP ID
rs74469108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:20746417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACAGCCTGTCACCAATCTTAAT[A/C]TTTAGAAAACAAGGCATTAAATTCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00320 PubMed Links

Gene Details

Gene
LINC00320
Gene Name
long intergenic non-protein coding RNA 320
There are no transcripts associated with this gene.

View Full Product Details