Product Details

SNP ID
rs77155265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35303272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCCTGAGGCTTGCCCCAAGCC[C/T]GACGTTGCTCTTTTACTCTCCCTTT
Phenotype
MIM: 604702 MIM: 604700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGXB4 PubMed Links

Gene Details

Gene
HMGXB4
Gene Name
HMG-box containing 4
There are no transcripts associated with this gene.

Gene
TOM1
Gene Name
target of myb1 membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135729.1 Intron NP_001129201.1
NM_001135730.1 Intron NP_001129202.1
NM_001135732.1 Intron NP_001129204.1
NM_005488.2 Intron NP_005479.1
XM_011529818.2 Intron XP_011528120.1
XM_011529820.2 Intron XP_011528122.1
XM_017028529.1 Intron XP_016884018.1
XM_017028530.1 Intron XP_016884019.1

View Full Product Details