Product Details
- SNP ID
-
rs79604243
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:35709552 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTCATTACATTTTCCCTGCTGCC[A/G]CCATTCAGCAACCATTTTTCCTGGG
- Phenotype
-
MIM: 607255
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APOL5
PubMed Links
Gene Details
- Gene
- APOL5
- Gene Name
- apolipoprotein L5
View Full Product Details