Product Details

SNP ID
rs77441740
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41933271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGAGGGAAAGCAGGCTAGAGTA[A/C]AGGGGCCCGCCAGGCAGCCCCTGAG
Phenotype
MIM: 610152 MIM: 606269
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CENPM PubMed Links

Gene Details

Gene
CENPM
Gene Name
centromere protein M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002876.2 Intron NP_001002876.1
NM_001110215.2 Intron NP_001103685.1
NM_001304370.1 Intron NP_001291299.1
NM_001304371.1 Intron NP_001291300.1
NM_001304372.1 Intron NP_001291301.1
NM_001304373.1 Intron NP_001291302.1
NM_024053.4 Intron NP_076958.1
XM_011530368.2 Intron XP_011528670.1
Gene
MIR378I
Gene Name
microRNA 378i
There are no transcripts associated with this gene.

Gene
TNFRSF13C
Gene Name
TNF receptor superfamily member 13C
There are no transcripts associated with this gene.

View Full Product Details