Product Details

SNP ID
rs75503117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43825231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTCCTTTGGTGGCCGACTCTCCA[A/C]CCACTGCATGCAAATGCTCCTGACT
Phenotype
MIM: 608359
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SULT4A1 PubMed Links

Gene Details

Gene
SULT4A1
Gene Name
sulfotransferase family 4A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014351.3 1346 UTR 3 NP_055166.1
XM_011530120.2 1346 Intron XP_011528422.1
XM_011530121.1 1346 UTR 3 XP_011528423.1

View Full Product Details