Product Details

SNP ID
rs76731590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23691017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTGGCTCTGTCTTGAAGATGCA[C/T]CTCTCGGAGCCTCGGTTGGCTCCTC
Phenotype
MIM: 612214
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUSBP11 PubMed Links

Gene Details

Gene
GUSBP11
Gene Name
glucuronidase, beta pseudogene 11
There are no transcripts associated with this gene.

Gene
RGL4
Gene Name
ral guanine nucleotide dissociation stimulator like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153615.1 157 UTR 5 NP_705843.1
XM_017028767.1 157 Intron XP_016884256.1

View Full Product Details