Product Details

SNP ID
rs76372697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102869998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGATTGTGTTGAGTTATGAGC[A/G]TTTCATTGTAAGGGGGATATTTCTA
Phenotype
MIM: 611507 MIM: 602963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CISD2 PubMed Links

Gene Details

Gene
CISD2
Gene Name
CDGSH iron sulfur domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008388.4 Intron NP_001008389.1
Gene
UBE2D3
Gene Name
ubiquitin conjugating enzyme E2 D3
There are no transcripts associated with this gene.

View Full Product Details