Product Details

SNP ID
rs79322681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144112055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTTCTTCTATTTGGGCACCA[C/T]AGGAACTGTGCATCTCAAACATTCT
Phenotype
MIM: 111300
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GYPA PubMed Links

Gene Details

Gene
GYPA
Gene Name
glycophorin A (MNS blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308187.1 Intron NP_001295116.1
NM_001308190.1 Intron NP_001295119.1
NM_002099.7 Intron NP_002090.4
XM_017008134.1 Intron XP_016863623.1
XM_017008135.1 Intron XP_016863624.1
XM_017008136.1 Intron XP_016863625.1

View Full Product Details