Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001029998.5 | 2901 | UTR 3 | NP_001025169.1 | ||
NM_001300842.2 | 2901 | UTR 3 | NP_001287771.1 | ||
NM_001317816.1 | 2901 | UTR 3 | NP_001304745.1 | ||
NM_001317817.1 | 2901 | Intron | NP_001304746.1 | ||
NM_001317818.1 | 2901 | Intron | NP_001304747.1 | ||
NM_032128.4 | 2901 | Intron | NP_115504.1 | ||
XM_011532311.2 | 2901 | UTR 3 | XP_011530613.1 | ||
XM_011532313.2 | 2901 | UTR 3 | XP_011530615.1 | ||
XM_011532314.2 | 2901 | UTR 3 | XP_011530616.1 | ||
XM_017008689.1 | 2901 | Intron | XP_016864178.1 | ||
XM_017008690.1 | 2901 | Intron | XP_016864179.1 | ||
XM_017008691.1 | 2901 | Intron | XP_016864180.1 | ||
XM_017008692.1 | 2901 | Intron | XP_016864181.1 |