Product Details

SNP ID
rs79250109
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:11857985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGATTCTGCCCCTTTGATAAGA[A/G]GTAGGAGGTGGGGAGAAGGTATTGT
Phenotype
MIM: 600295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPPB PubMed Links

Gene Details

Gene
NPPB
Gene Name
natriuretic peptide B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002521.2 Intron NP_002512.1

View Full Product Details