Product Details

SNP ID
rs77839787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:131117780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGATGTGATACTGTCTACAACA[C/T]TCCTTCCTTTTTGTTTCGCCTGTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHB2 PubMed Links

Gene Details

Gene
PLEKHB2
Gene Name
pleckstrin homology domain containing B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100623.1 Intron NP_001094093.1
NM_001267062.1 Intron NP_001253991.1
NM_001267063.1 Intron NP_001253992.1
NM_001267064.1 Intron NP_001253993.1
NM_001267065.1 Intron NP_001253994.1
NM_001267066.1 Intron NP_001253995.1
NM_001267067.1 Intron NP_001253996.1
NM_001267068.1 Intron NP_001253997.1
NM_001309448.1 Intron NP_001296377.1
NM_001309450.1 Intron NP_001296379.1
NM_001309451.1 Intron NP_001296380.1
NM_001309452.1 Intron NP_001296381.1
NM_017958.2 Intron NP_060428.2
XM_011511385.2 Intron XP_011509687.1
XM_011511386.2 Intron XP_011509688.1
XM_017004408.1 Intron XP_016859897.1
XM_017004409.1 Intron XP_016859898.1
XM_017004410.1 Intron XP_016859899.1
XM_017004411.1 Intron XP_016859900.1
XM_017004412.1 Intron XP_016859901.1
XM_017004413.1 Intron XP_016859902.1
Gene
POTEI
Gene Name
POTE ankyrin domain family member I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277406.1 Intron NP_001264335.1
XM_011511670.2 Intron XP_011509972.1
XM_017004732.1 Intron XP_016860221.1
XM_017004733.1 Intron XP_016860222.1
XM_017004734.1 Intron XP_016860223.1
XM_017004735.1 Intron XP_016860224.1

View Full Product Details