Product Details

SNP ID
rs75495651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108787461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTGAATAGAAATAACTTGTGC[A/G]TTTTTCCTAAGCATAAGGACGTTGT
Phenotype
MIM: 601181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC138 PubMed Links

Gene Details

Gene
CCDC138
Gene Name
coiled-coil domain containing 138
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303105.1 Intron NP_001290034.1
NM_001303106.1 Intron NP_001290035.1
NM_144978.2 Intron NP_659415.1
XM_006712344.2 Intron XP_006712407.1
XM_006712345.3 Intron XP_006712408.1
XM_011510757.2 Intron XP_011509059.1
XM_011510758.2 Intron XP_011509060.1
XM_011510760.2 Intron XP_011509062.1
XM_011510761.2 Intron XP_011509063.1
XM_011510762.2 Intron XP_011509064.1
XM_017003492.1 Intron XP_016858981.1
XM_017003493.1 Intron XP_016858982.1
XM_017003494.1 Intron XP_016858983.1
XM_017003495.1 Intron XP_016858984.1
XM_017003496.1 Intron XP_016858985.1
XM_017003497.1 Intron XP_016858986.1
XM_017003498.1 Intron XP_016858987.1
XM_017003499.1 Intron XP_016858988.1
XM_017003500.1 Intron XP_016858989.1
XM_017003501.1 Intron XP_016858990.1
XM_017003502.1 Intron XP_016858991.1
XM_017003503.1 Intron XP_016858992.1
XM_017003504.1 Intron XP_016858993.1
Gene
RANBP2
Gene Name
RAN binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006267.4 Intron NP_006258.3
XM_005264002.2 Intron XP_005264059.1
XM_005264003.2 Intron XP_005264060.1
XM_005264004.2 Intron XP_005264061.1
XM_005264005.4 Intron XP_005264062.1
XM_005264007.2 Intron XP_005264064.1
XM_011511575.2 Intron XP_011509877.1
XM_011511576.2 Intron XP_011509878.1
XM_011511578.2 Intron XP_011509880.1
XM_017004623.1 Intron XP_016860112.1
XM_017004624.1 Intron XP_016860113.1
XM_017004625.1 Intron XP_016860114.1

View Full Product Details