Product Details

SNP ID
rs74571554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:98622107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTAAGAAATCTTACATCTTAGAA[C/T]CCTAGAAATGGGTCCTCAGCTTTCT
Phenotype
MIM: 604623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGAT4A PubMed Links

Gene Details

Gene
MGAT4A
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160154.1 5381 Intron NP_001153626.1
NM_012214.2 5381 UTR 3 NP_036346.1
Gene
UNC50
Gene Name
unc-50 inner nuclear membrane RNA binding protein
There are no transcripts associated with this gene.

View Full Product Details