Product Details

SNP ID
rs74633464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99337979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATGCCTCTTATACCTGTGAGT[A/G]CTTTGGGAGGGAAGGGAGCTTGAAT
Phenotype
MIM: 606086 MIM: 612564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF5B PubMed Links

Gene Details

Gene
EIF5B
Gene Name
eukaryotic translation initiation factor 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015904.3 Intron NP_056988.3
Gene
TXNDC9
Gene Name
thioredoxin domain containing 9
There are no transcripts associated with this gene.

View Full Product Details