Product Details

SNP ID
rs76266505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43643566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGATTCTATATTACGCTAAAT[A/G]CTGGGAAAATCAATGTGGTTATATT
Phenotype
MIM: 612723
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHH2 PubMed Links

Gene Details

Gene
PLEKHH2
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172069.3 Intron NP_742066.2
XM_017003351.1 Intron XP_016858840.1
XM_017003352.1 Intron XP_016858841.1
XM_017003353.1 Intron XP_016858842.1

View Full Product Details