Product Details
- SNP ID
-
rs77838429
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:49920252 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAATTAACAGAGAATGATTTTTTA[A/G]GTGGGTAGTAATACAGAATCAAAAA
- Phenotype
-
MIM: 600565
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NRXN1
PubMed Links
Gene Details
- Gene
- NRXN1
- Gene Name
- neurexin 1
View Full Product Details