Product Details

SNP ID
rs77201768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64187802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGTTCCTTTCTTCTGCAGGAA[G/T]GCCTTCTCATTAGCTCTTAGTTCAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00309 PubMed Links

Gene Details

Gene
LINC00309
Gene Name
long intergenic non-protein coding RNA 309
There are no transcripts associated with this gene.

View Full Product Details