Product Details
- SNP ID
-
rs79530167
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:64455312 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCAGCCCCCCAAAAAAGAGCCCA[G/T]GGAAAGCCAATCTGTGTACTTCTAT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LGALSL
PubMed Links
Gene Details
- Gene
- LGALSL
- Gene Name
- galectin like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014181.2 |
|
Intron |
|
|
NP_054900.2 |
View Full Product Details