Product Details

SNP ID
rs76623705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:67397667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCCTGTGCAGTAAAAGTAACCC[C/G]GAGGGTGAGACGTCGGCAGCGCGGC
Phenotype
MIM: 613196
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ETAA1 PubMed Links

Gene Details

Gene
ETAA1
Gene Name
Ewing tumor associated antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019002.3 351 Silent Mutation CCC,CCG P73P NP_061875.2
XM_017004376.1 351 Silent Mutation CCC,CCG P73P XP_016859865.1
XM_017004377.1 351 Silent Mutation CCC,CCG P73P XP_016859866.1

View Full Product Details