Product Details

SNP ID
rs80280553
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74425916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAACTTTAATGGTTGATGTGGGA[A/G]TCACAAGGGAGGTATGTTTGCTCCA
Phenotype
MIM: 602288
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C2orf81 PubMed Links

Gene Details

Gene
C2orf81
Gene Name
chromosome 2 open reading frame 81
There are no transcripts associated with this gene.

Gene
RTKN
Gene Name
rhotekin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015055.1 2755 UTR 3 NP_001015055.1
NM_001015056.1 2755 UTR 3 NP_001015056.1
NM_033046.2 2755 UTR 3 NP_149035.1
XM_005264478.2 2755 Intron XP_005264535.1
XM_005264479.4 2755 Intron XP_005264536.1
XM_017004635.1 2755 UTR 3 XP_016860124.1
XM_017004636.1 2755 UTR 3 XP_016860125.1
XM_017004637.1 2755 UTR 3 XP_016860126.1
XM_017004638.1 2755 UTR 3 XP_016860127.1
XM_017004639.1 2755 Intron XP_016860128.1
XM_017004640.1 2755 Intron XP_016860129.1
Gene
WDR54
Gene Name
WD repeat domain 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320823.1 2755 Intron NP_001307752.1
NM_001320824.1 2755 Intron NP_001307753.1
NM_001320825.1 2755 Intron NP_001307754.1
NM_032118.3 2755 Intron NP_115494.1
XM_017005064.1 2755 Intron XP_016860553.1

View Full Product Details