Product Details
- SNP ID
-
rs77996146
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:11282394 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATGTGACTGCTTGTTCCCTGGTT[A/G]ACACACTGAAGAAGAAATCATGTTG
- Phenotype
-
MIM: 608760
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATG7
PubMed Links
Gene Details
- Gene
- ATG7
- Gene Name
- autophagy related 7
View Full Product Details