Product Details

SNP ID
rs75232354
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:19159558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCATTGTTGCATGAAAACAGGG[A/C]CAAGATTGGTTGGCAGATCTCCAGT
Phenotype
MIM: 608260
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
KCNH8 PubMed Links

Gene Details

Gene
KCNH8
Gene Name
potassium voltage-gated channel subfamily H member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144633.2 Intron NP_653234.2
XM_011533347.2 Intron XP_011531649.1
XM_017005698.1 Intron XP_016861187.1
XM_017005699.1 Intron XP_016861188.1
XM_017005700.1 Intron XP_016861189.1
XM_017005701.1 Intron XP_016861190.1
XM_017005702.1 Intron XP_016861191.1

View Full Product Details