Product Details

SNP ID
rs78999570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32257843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGCTTAAGTGAGAATCTCTGT[A/G]CCTCGGCAGTGAATACAGGTTTTCA
Phenotype
MIM: 607891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMTM8 PubMed Links

Gene Details

Gene
CMTM8
Gene Name
CKLF like MARVEL transmembrane domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320308.1 Intron NP_001307237.1
NM_178868.4 Intron NP_849199.2
XM_011533416.2 Intron XP_011531718.1
XM_017005779.1 Intron XP_016861268.1
XM_017005780.1 Intron XP_016861269.1

View Full Product Details