Product Details
- SNP ID
-
rs77773073
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:44877917 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAATGGTCTACATGTCCATCCAG[A/G]GGGATAATAGCTACTAAAAAGTAAC
- Phenotype
-
MIM: 600585
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TGM4
PubMed Links
Gene Details
- Gene
- TGM4
- Gene Name
- transglutaminase 4
View Full Product Details