Product Details

SNP ID
rs77813979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49153792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGTCAAAGTGACAACTGTAGA[C/T]ATGCCCATGGCACTCACATTCTGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC71 PubMed Links

Gene Details

Gene
CCDC71
Gene Name
coiled-coil domain containing 71
There are no transcripts associated with this gene.

Gene
LAMB2P1
Gene Name
laminin subunit beta 2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details