Product Details

SNP ID
rs79096402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51679773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTATAGAGGAAGGTTCCTGAAA[A/G]GTTTCTAAGTAAACTCACCTTTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX264 PubMed Links

Gene Details

Gene
TEX264
Gene Name
testis expressed 264
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129884.2 Intron NP_001123356.1
NM_001243725.1 Intron NP_001230654.1
NM_001243726.1 Intron NP_001230655.1
NM_001243727.2 Intron NP_001230656.1
NM_001278195.1 Intron NP_001265124.1
NM_015926.5 Intron NP_057010.1
XM_006713195.3 Intron XP_006713258.1
XM_006713197.3 Intron XP_006713260.1
XM_006713198.2 Intron XP_006713261.1
XM_011533805.2 Intron XP_011532107.1
XM_017006573.1 Intron XP_016862062.1
XM_017006574.1 Intron XP_016862063.1

View Full Product Details