Product Details
- SNP ID
-
rs75073117
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:8622003 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGAAGTTTGAGGCCACAAATTGCT[C/T]AAGACATCAATTGCCATCAAGTGGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SSUH2
PubMed Links
Gene Details
- Gene
- SSUH2
- Gene Name
- ssu-2 homolog (C. elegans)
View Full Product Details