Product Details

SNP ID
rs79463547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177503803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGCTTTATTATCTGTGAGTCT[C/G]CACACTATTCATGAGGAGGGCAGGG
Phenotype
MIM: 608170 MIM: 611435 MIM: 605903
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX41 PubMed Links

Gene Details

Gene
DDX41
Gene Name
DEAD-box helicase 41
There are no transcripts associated with this gene.

Gene
DOK3
Gene Name
docking protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144875.1 1587 Intron NP_001138347.1
NM_001144876.1 1587 Intron NP_001138348.1
NM_001308235.1 1587 UTR 3 NP_001295164.1
NM_001308236.1 1587 UTR 3 NP_001295165.1
NM_024872.2 1587 UTR 3 NP_079148.2
XM_005265985.3 1587 UTR 3 XP_005266042.1
XM_005265987.1 1587 Intron XP_005266044.1
XM_011534654.2 1587 UTR 3 XP_011532956.1
XM_011534655.2 1587 UTR 3 XP_011532957.1
XM_011534656.2 1587 UTR 3 XP_011532958.1
XM_011534657.2 1587 UTR 3 XP_011532959.1
XM_017009883.1 1587 UTR 3 XP_016865372.1
XM_017009884.1 1587 Intron XP_016865373.1
Gene
PDLIM7
Gene Name
PDZ and LIM domain 7
There are no transcripts associated with this gene.

View Full Product Details