Product Details

SNP ID
rs75843251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178941409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTGCGGGTTGTGGTCCATTCTG[A/G]AGGACGCTGATCGAATGCCCCAAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP2 PubMed Links

Gene Details

Gene
ZFP2
Gene Name
ZFP2 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZNF454
Gene Name
zinc finger protein 454
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178089.2 219 UTR 5 NP_001171560.1
NM_001178090.2 219 Intron NP_001171561.1
NM_001323306.1 219 UTR 5 NP_001310235.1
NM_001323307.1 219 UTR 5 NP_001310236.1
NM_001323308.1 219 UTR 5 NP_001310237.1
NM_001323309.1 219 Intron NP_001310238.1
NM_001323310.1 219 Intron NP_001310239.1
NM_182594.3 219 Intron NP_872400.2

View Full Product Details