Product Details

SNP ID
rs79131526
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160349382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGTCCGGATGCGGTACTGGCCG[G/T]TGTGCACCAGGCCGATGGCCAGGTG
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
C1QTNF2 PubMed Links

Gene Details

Gene
C1QTNF2
Gene Name
C1q and tumor necrosis factor related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031908.4 806 Missense Mutation AAC,ACC N260T NP_114114.2
XM_011534425.2 806 Missense Mutation AAC,ACC N299T XP_011532727.1
XM_011534426.2 806 Missense Mutation AAC,ACC N231T XP_011532728.1
XM_011534427.2 806 UTR 3 XP_011532729.1
XM_017009013.1 806 Missense Mutation AAC,ACC N192T XP_016864502.1
Gene
CCNJL
Gene Name
cyclin J like
There are no transcripts associated with this gene.

View Full Product Details