Product Details

SNP ID
rs76989603
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:31138048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTTGTTTCCCTATAGCACTTAA[C/T]ATCTCTGATAGATGACACAGGCTTG
Phenotype
MIM: 610703
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HSPH1 PubMed Links

Gene Details

Gene
HSPH1
Gene Name
heat shock protein family H (Hsp110) member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286503.1 Intron NP_001273432.1
NM_001286504.1 Intron NP_001273433.1
NM_001286505.1 Intron NP_001273434.1
NM_006644.3 Intron NP_006635.2
XM_005266236.1 Intron XP_005266293.1
XM_011534887.2 Intron XP_011533189.1
XM_011534888.1 Intron XP_011533190.1
XM_017020361.1 Intron XP_016875850.1
XM_017020362.1 Intron XP_016875851.1
XM_017020363.1 Intron XP_016875852.1
XM_017020364.1 Intron XP_016875853.1

View Full Product Details