Product Details
- SNP ID
-
rs80229802
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:20709638 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCCTTAGAACGTGGGTTCTGTGTG[C/T]GTCTTATGAGGGTCTCTGGAGGCTG
- Phenotype
-
MIM: 607587
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL17D
PubMed Links
Gene Details
- Gene
- IL17D
- Gene Name
- interleukin 17D
View Full Product Details