Product Details

SNP ID
rs78307699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:48305549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAAGCATACTCTAATGTAGGGT[A/G]ACCAGGAATATTGACGTTGTGGCCA
Phenotype
MIM: 614041
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00441 PubMed Links

Gene Details

Gene
LINC00441
Gene Name
long intergenic non-protein coding RNA 441
There are no transcripts associated with this gene.

Gene
RB1
Gene Name
RB transcriptional corepressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000321.2 Intron NP_000312.2
XM_011535171.2 Intron XP_011533473.1

View Full Product Details