Product Details

SNP ID
rs78557143
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62044626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTGCCAAACTGATTTCTGACT[C/T]TTAAATTGTATTTATTCACTCCCCA
Phenotype
MIM: 606097
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PIGN PubMed Links
Additional Information
For this assay, SNP(s) [rs34860757] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PIGN
Gene Name
phosphatidylinositol glycan anchor biosynthesis class N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012327.5 3630 UTR 3 NP_036459.1
NM_176787.4 3630 UTR 3 NP_789744.1
XM_011525889.1 3630 UTR 3 XP_011524191.1
XM_011525890.1 3630 UTR 3 XP_011524192.1
XM_011525891.1 3630 UTR 3 XP_011524193.1
XM_011525892.1 3630 UTR 3 XP_011524194.1
XM_011525893.1 3630 UTR 3 XP_011524195.1
XM_011525894.1 3630 UTR 3 XP_011524196.1
XM_011525895.1 3630 UTR 3 XP_011524197.1
XM_011525896.1 3630 UTR 3 XP_011524198.1
XM_011525898.1 3630 Intron XP_011524200.1
XM_017025685.1 3630 UTR 3 XP_016881174.1
XM_017025686.1 3630 UTR 3 XP_016881175.1

View Full Product Details