Product Details

SNP ID
rs79354666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110424943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGCCCACTATTATTAGTTGTG[A/G]GAAGTAATTTGCTTGACTTGCTTTC
Phenotype
MIM: 124450 MIM: 608276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDO PubMed Links

Gene Details

Gene
DDO
Gene Name
D-aspartate oxidase
There are no transcripts associated with this gene.

Gene
SLC22A16
Gene Name
solute carrier family 22 member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033125.3 1235 Missense Mutation CCC,CTC P555L NP_149116.2
XM_005267184.3 1235 Missense Mutation CCC,CTC P385L XP_005267241.1
XM_011536204.2 1235 Missense Mutation CCC,CTC P512L XP_011534506.1
XM_011536205.2 1235 UTR 3 XP_011534507.1
XM_011536206.2 1235 Missense Mutation CCC,CTC P461L XP_011534508.1
XM_011536207.2 1235 Missense Mutation CCC,CTC P437L XP_011534509.1
XM_011536208.2 1235 Missense Mutation CCC,CTC P425L XP_011534510.1
XM_011536209.2 1235 Missense Mutation CCC,CTC P420L XP_011534511.1
XM_011536210.1 1235 Intron XP_011534512.1
XM_011536211.2 1235 Missense Mutation CCC,CTC P395L XP_011534513.1
XM_011536212.2 1235 Missense Mutation CCC,CTC P325L XP_011534514.1

View Full Product Details