Product Details

SNP ID
rs77068590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110426203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACATTCCAAAGGAATTTTAAAGA[C/T]GTTTTCTGGTCTCTCAAGACACGGC
Phenotype
MIM: 608276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC22A16 PubMed Links

Gene Details

Gene
SLC22A16
Gene Name
solute carrier family 22 member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033125.3 Intron NP_149116.2
XM_005267184.3 Intron XP_005267241.1
XM_011536204.2 Intron XP_011534506.1
XM_011536205.2 Intron XP_011534507.1
XM_011536206.2 Intron XP_011534508.1
XM_011536207.2 Intron XP_011534509.1
XM_011536208.2 Intron XP_011534510.1
XM_011536209.2 Intron XP_011534511.1
XM_011536210.1 Intron XP_011534512.1
XM_011536211.2 Intron XP_011534513.1
XM_011536212.2 Intron XP_011534514.1

View Full Product Details