Product Details

SNP ID
rs77865145
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:134961207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATCAGAGTAACAAAATAATGTA[C/G]TCCCTTTATGGACTATAAGTAACTT
Phenotype
MIM: 612450
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
HBS1L PubMed Links

Gene Details

Gene
HBS1L
Gene Name
HBS1 like translational GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145158.1 6334 UTR 3 NP_001138630.1
NM_001145207.1 6334 Intron NP_001138679.1
NM_006620.3 6334 UTR 3 NP_006611.1
XM_017010192.1 6334 Intron XP_016865681.1

View Full Product Details